Order Entry
Ireland
ContactUsLinkComponent
Germanium ≥99.9999% (metals basis), polycrystalline rods, Ø 6.35 mm (0.25 in)
Germanium ≥99.9999% (metals basis), polycrystalline rods, Ø 6.35 mm (0.25 in)
Catalog # 42951.BS
CAS Number:  
Germanium ≥99.9999% (metals basis), polycrystalline rods, Ø 6.35 mm (0.25 in)
Catalog # 42951.BS
CAS Number:  

Specifications

  • Pk:
    10 cm
  • Seq:
    7600

Specifications

About this item

 :
 :72.61 g/mol
Merck Index:12,04415
Boiling Pt:2830 °C (1013 hPa)
Melting Pt:937,2 °C
Density:5,323 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-164-3
CAS Number:7440-56-4
Formula:Ge
MDL Number:MFCD00085310