Germanium ≥99.9999% (metals basis) (N type ≥ 95%), ingot, zone refined 95% N type minimum
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-56-4
:
Formula:Ge
MDL Number:MFCD00085310
:72.61 g/mol
Merck Index:12,04415
Boiling Pt:2830 °C (1013 hPa)
Melting Pt:937,2 °C
Density:5,323 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-164-3
Related Information
Frequently Bought Together