Aluminium for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7429-90-5
:
Formula:Al
MDL Number:MFCD00134029
:26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2,7 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:231-072-3
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:660 °C
- Molar Mass:26.98 g/mol
- Boiling Point:2467 °C
- Density:2.70 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together