Order Entry
Ireland
ContactUsLinkComponent
Aluminium ≥99.9995% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Aluminium ≥99.9995% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Catalog # 43392.30
CAS Number:  
Aluminium ≥99.9995% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Catalog # 43392.30
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2,7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-072-3
CAS Number:7429-90-5
Formula:Al
MDL Number:MFCD00134029