Aluminium ≥99.9995% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
: Aluminum
:7429-90-5
:
:Al
:MFCD00134029
:26.98 g/mol
:13,00321
:2327 °C (1013 hPa)
:660 °C
:2,7 g/cm³ (20 °C)
:Ambient
:231-072-3
- Environmentally Preferable:
Frequently Bought Together