Aluminium ≥99.99% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Aluminum
CAS Number:7429-90-5
:
Formula:Al
MDL Number:MFCD00134029
:26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2,7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-072-3
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together