Aluminium ≥99% (metals basis), foil [Si & Fe (combined) ∼ 1%], annealed, Thickness: 1.0 mm (0.04 in)
Catalog # 14192.RV
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:7825
- Environmentally Preferable:
- Pk Info:45×60 cm
Specifications
About this item
Synonyms: Aluminum
:
:26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2,7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-072-3
CAS Number:7429-90-5
Formula:Al
MDL Number:MFCD00134029