Order Entry
Ireland
ContactUsLinkComponent
Ammonium hydrogen carbonate, LiChropur™ for LC-MS, Supelco®
Ammonium hydrogen carbonate, LiChropur™ for LC-MS, Supelco®
Catalog # 5.33005.0050
Supplier:  Merck
CAS Number:  
Ammonium hydrogen carbonate, LiChropur™ for LC-MS, Supelco®
Catalog # 5.33005.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • pH value (50 g/l, H₂O, 20 °C):
    8
  • Vapor pressure:
    67 hPa (20 °C)
  • Solubility:
    220 g/l

Specification Test Results

About this item

 :
 :79.06 g/mol
Melting Pt:106 °C
EINECS:213-911-5
 :GHS07
Storage Temperature:Ambient
CAS Number:1066-33-7
Formula:NH₄HCO₃
MDL Number:MFCD00012138