Order Entry
Ireland
ContactUsLinkComponent
Spectrophotometer cuvette
Spectrophotometer cuvette
Catalog # MINO1870-716
Supplier:  MINOLTA BUSINESS
Spectrophotometer cuvette
Catalog # MINO1870-716
Supplier:  MINOLTA BUSINESS
Specifications
  • Path length:
    10 mm
  • Window material:
    PMMA
  • Description:
    Rectangular disposal cells
  • Height:
    50 mm
  • Pk:
    100
Specifications
About this item
Rectangular disposal cell suitable for transmittance measurements of water based transparent liquids.
Recommended for applications with liquids of normal density.