Order Entry
Ireland
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Imidacloprid
  37894-100MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Imidacloprid
  37894-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    138261-41-3
  • Description:
    Imidacloprid
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H301 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00468059
  • M-Clarity quality level:
    MQ100
  • Melting point:
    140...146 °C
  • Formula:
    C₉H₁₀CLN₅O₂
  • Molecular weight:
    255,66
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS07, GHS09
 : P264 - P273 - P301 + P310 - P391 - P405 - P501