Z-Leu-Leu-Leu-B(OH)2
ENZOBMLPI1090100
:
- Pk:100 µG
- Seq:02
- Protein/peptide name:Z-Leu-Leu-Leu-B(OH)2
- Purity:99%
Proteasome inhibitor
A potent cell-permeable proteasome inhibitor (Ki=0,023 nM).
Proteasome inhibitor
A potent cell-permeable proteasome inhibitor (Ki=0,023 nM).