Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
AIP
AIP
AIP
Catalog # ENZOBMLP2120500
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
AIP
Catalog # ENZOBMLP2120500
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
0,5 mg
Seq:
02
Protein/peptide name:
AIP
Molecular weight:
1708,1
Specifications
About this item
Product Details
CaM kinase inhibitor
Product Details