Order Entry
Ireland
ContactUsLinkComponent
 
G-type-iODN
  ENZOALX746252C100
 :  
G-type-iODN
  ENZOALX746252C100
 :  

 

  • Pk:
    100 µG
  • Seq:
    02
  • Reconstitution instructions:
    For a 100 µM stock solution, dissolve the total vial content in 150 µl endotoxin-free water (included) or PBS (to order separately). To obtain optimal dissolving we recommend the following procedure:
    - Add 50% of the solvent and let dissolve for 10 min.
  • Endotoxin-free:
    Yes
  • Protein/peptide name:
    G-type-iODN
  • Formulation:
    Lyophilised. Sterile.

 

 

TLR9 inhibitor

Inhibitory iODN oligodeoxynucleotide with phosphorothioate backbone. Potent sequence of an inhibitory ODN based on poly-G class I sequences targeting TLR9, but without additional effect on TLR7 and TLR8 signaling, for in vivo use in rodents (50-150 µg per injection). For active CpG compounds see e.g. ODN 1668 (Prod. No. ALX-746-001).

Synthetic.

 : Quantity: ~15 nmol (100 µg). Sufficient for ~100 cellular inhibition assays. Working concentration depends upon concentration, type and MW of stimulating CpG ODN, the cellular system and read-out used. Titrate at 3-5 fold molar excess for inhibition.