Z-VAD-FMK
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Caspase inhibitor
Irreversible, cell permeable broad-spectrum caspase inhibitor.
Specifications
- Protein/peptide name:Z-VAD-FMK
- Purity:>92% (HPLC)