Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Proteins and Peptides
PKI-tide
PKI-tide
Print
PKI-tide
Catalog #
PKI-tide
Catalog #
Supplier:
CAS Number:
Expand image
Return to Top
PKI-tide
Add to cart
CAS Number:
Return Policy
Product Details & Documents
Specific inhibitor of cAMP-dependent protein kinase (PKA).
Specific PKA inhibitor
Soluble in water.