Order Entry
Ireland
Orders LinkContactUsLinkComponent
Human recombinant IL-8

Human recombinant IL-8

25 µG
Catalog # ENZOENZPRT1850025
Supplier:   Enzo Life Sciences
Human recombinant IL-8
Catalog # ENZOENZPRT1850025
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant IL-8

25 µG

CAS Number:  
Price
314,00 €
Log in for availability
1 * 25 µG
Product Details & Documents

Produced in E. coli.

Specifications

Specifications

  • Pk:
    25 µG
  • Cat. No.:
    ENZOENZPRT1850025
  • Protein/peptide type:
    Recombinant
  • Purification:
    Sterile filtered through a 0,2 micron filter.
  • Protein/peptide name:
    IL-8
  • Formulation:
    Lyophilized with no additives.
  • Protein synonyms:
    MDNCF|Neutrophil activating factor|Interleukin-8|NAP-1|Monocyte-derived neutrophil chemotactic factor|NAF|CXCL8
  • Purity:
    >98% (SDS and HPLC)
  • Species:
    Human
  • Molecular weight:
    ~8,4 kDa

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.