Order Entry
Ireland
ContactUsLinkComponent
Human recombinant vitronectin
Human recombinant vitronectin
Catalog # ENZOENZPRT1790500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant vitronectin
Catalog # ENZOENZPRT1790500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    500 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein synonyms:
    Serum-spreading factor|VTN|V75
  • Protein/peptide name:
    Vitronectin
  • Purity:
    >95% (SDS and HPLC)
  • Molecular weight:
    ~52,3 kDa
  • Formulation:
    Lyophilized from 20 mM sodium phosphate, pH 7,5.
  • Purification:
    Sterile filtered through a 0,2 micron filter.

Specifications

About this item

Produced in HEK 293 cells.