Order Entry
Ireland
ContactUsLinkComponent
 
Human recombinant Follistatin
 
Human recombinant Follistatin

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Biological activity:
    The activity is determined by the dose-dependent proliferation of mouse BALB/c 3T3 cells which is typically observed at concentations <0,1 ng/ml.
  • Protein synonyms:
    FSH-suppressing protein|FS|Activin-binding protein|FSP
  • Protein/peptide name:
    Follistatin
  • Purity:
    >95% (HPLC, Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
  • Molecular weight:
    ~31,7kDa
  • Formulation:
    Lyophilized from 10 mM Na2PO4, pH 7,5. Sterile filtered.

 

Frequently Bought Together