Recombinant polyubiquitin chains (Ub2-7) (linear)
Catalog # ENZOBMLUW10100100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Seq:02
- Protein/peptide name:Polyubiquitin Chains (Ub2-7) (linear)
- Purity:>95% (SDS-PAGE)
- Molecular weight:Ub2: ~17,1 kDa; Ub3: ~25,6 kDa; Ub4: ~34,2 kDa; Ub5: ~42,7 kDa; Ub6: ~51,3 kDa; Ub7: ~59,8 kDa
- Formulation:Liquid. In 50 mM TRIS-HCl, pH 8,0, containing 50 mM sodium chloride.
- Purification:Purified by multi-step chromatography
Specifications
About this item
Produced in E. coli.
Linear Polyubiquitin chains Ub2-Ub7