Human recombinant HDAC3, HIS tag
ENZOBMLSE5070050
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:HDAC3
Highly active
Produced in insect cells.
Highly active
Produced in insect cells.