Order Entry
Ireland
ContactUsLinkComponent
Mouse recombinant Leptin (from E. coli)
Mouse recombinant Leptin (from E. coli)
Catalog # ENZOBMLSE1600001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Mouse recombinant Leptin (from E. coli)
Catalog # ENZOBMLSE1600001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Seq:
    02
  • Protein/peptide name:
    Leptin
  • Purity:
    >95% (SDS-PAGE)
  • Formulation:
    Lyophilised

Specifications

About this item

Produced in E. coli.