Human recombinant HDAC8 (from E. coli)
Catalog # ENZOBMLSE1450100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 Vial
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Biological activity:Like other class I HDACs, HDAC8 exhibits trichostatin A-inhibitable histone deacetylase activity and can mediate transcription repression.
- Protein/peptide name:HDAC8
- Formulation:Liquid. In 10 mM TRIS, pH 7,5, containing 100 mM NaCl, 3 mM MgCl2 and 10% glycerol.
Specifications
About this item
Produced in E. coli.