IKK substrate
Catalog # ENZOBMLP2260001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:IKK substrate
- Purity:95%
Specifications
About this item
This peptide, KKKKERLLDDRHDSGLDSMKDEE, was derived from the IkappaBalpha sequence.
The four lysines on the amino-terminus allow this peptide to be used in phosphocellulose kinase assays for IKK activity.