Abz-SDK(Dnp)-P
ENZOBMLP1620001
:
- Pk:1 mg
- Seq:02
- Protein/peptide name:Abz-SDK(Dnp)-P
- Purity:97%
ACE substrate
Fluorogenic substrate for angiotensin converting enzyme-1 (ACE1) N-domain. The somatic form of ACE1 is a zinc-dependent peptidyl dipeptidase comprising two catalytic domains referred to as the N- and C-domains. The testicular form, due to a downstream start site, contains only the C-domain. Abz fluorescence is quenched by the Dnp group until cleavage (at Asp-Lys) separates them. Ex: 320 nm, Em: 420 nm. This quenched substrate is useful for assay of cell culture ACE1 N-domain activity, inhibitor screening and kinetic analysis.