Human;Mouse recombinant Fc(Human):Dectin-2
Catalog # ENZOALX522126C050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:HumanMouse
- Seq:02
- Reconstitution instructions:Reconstitute with 50 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum.
- Protein/peptide name:Fc(human):Dectin-2
- Purity:>95% (SDS-PAGE)
- Concentration:1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Specifications
About this item
Produced in HEK 293 cells. The extracellular domain of dectin-2 (aa 44-209) is fused at the N-terminus to the Fc portion of human IgG1.