Order Entry
Ireland
ContactUsLinkComponent
Human recombinant TNF-R1:Fc
Human recombinant TNF-R1:Fc
Catalog # ENZOALX522013C050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant TNF-R1:Fc
Catalog # ENZOALX522013C050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Biological activity:
    Binds to TNF-R1 and triggers various biological effects (including cell death). rhTNF-R1:Fc fusion protein blocks TNF-α-mediated lysis with a inhibitory activity in a concentration range of 10 - 50 ng/ml.
  • Reconstitution instructions:
    Reconstitute with 50 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or carrier protein.
  • Protein/peptide name:
    TNF-R1:Fc
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

Specifications

About this item

Produced in HEK 293 cells. The extracellular domain of human TNF-R1 (aa 1-211) is fused to the Fc portion of human IgG1.