Order Entry
Ireland
ContactUsLinkComponent
Microcystis aeruginosa Microcystin LY
Microcystis aeruginosa Microcystin LY
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
Microcystis aeruginosa Microcystin LY
Supplier:  ENZO LIFE SCIENCES

Specifications

  • CAS No.:
    123304-10-9
  • Species:
    Microcystis aeruginosa
  • Protein/peptide name:
    Microcystin LY
  • Purity:
    >95% (HPLC)

Specifications

Frequently Bought Together