0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Proteins and Peptides
HDAC Substrate (fluorogenic)
HDAC Substrate (fluorogenic)
:
ENZO LIFE SCIENCES
HDAC Substrate (fluorogenic)
:
ENZO LIFE SCIENCES
Protein/peptide name:
HDAC Substrate (fluorogenic)
Purity:
>99%
Frequently Bought Together
VWR® Premium Line, Serological Pipettes
: VWR Collection
VWR®, Test Tubes without Rim
: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Serological Pipette, Standard Line
: VWR Collection
Serological pipettes, Stripette™
: Corning
VWR®, Aspirating Pipettes
: VWR Collection
VWR®, Sampling Scoops
: VWR Collection
VWR® Ultra-High Performance Centrifuge Tubes
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR® Cryoboxes and Dividers
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Aluminium tool for vial transfer, CryoHolder
: PHARSOL