Ac-LEVD-pNA
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Caspase-4 substrate
Chromogenic substrate for caspase-4.
Specifications
- Protein/peptide name:Ac-LEVD-pNA
- Purity:>99% (HPLC)
Specifications
Frequently Bought Together
Caspase-4 substrate
Chromogenic substrate for caspase-4.