Order Entry
Ireland
ContactUsLinkComponent
Mca-YVADAPK(Dnp)-OH
Mca-YVADAPK(Dnp)-OH
Catalog # ENZOALX260023M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Mca-YVADAPK(Dnp)-OH
Catalog # ENZOALX260023M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    Mca-YVADAPK(Dnp)-OH
  • Purity:
    >97% (HPLC)

Specifications

About this item

Substrate for caspase-1 and -4

Specific, highly fluorescent substrate for caspase-1. Also substrate for caspase-4.