Order Entry
Ireland
ContactUsLinkComponent
Substance P
Substance P
Supplier:  ENZO LIFE SCIENCES
Certificates
Substance P
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide name:
    Substance P
  • Purity:
    >97% (HPLC)
  • Formulation:
    Lyophilised

Specifications

Frequently Bought Together