Order Entry
Ireland
ContactUsLinkComponent
Human recombinant p23 (from E. coli)
Human recombinant p23 (from E. coli)
Supplier:  ENZO LIFE SCIENCES
Certificates
Human recombinant p23 (from E. coli)
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    p23

Specifications

Frequently Bought Together