Order Entry
Ireland
ContactUsLinkComponent
 
Bovine native alpha-Crystallin (from Eye lens)
  ENZOADISPP225J
 :  
Bovine native alpha-Crystallin (from Eye lens)
  ENZOADISPP225J
 :  

 

  • Pk:
    1 mg
  • Protein/peptide type:
    Native
  • Source:
    Eye lens
  • Species:
    Bovine
  • Seq:
    02
  • Protein synonyms:
    α-Crystallin
  • Protein/peptide name:
    alpha-Crystallin
  • Formulation:
    Liquid, in PBS, pH 7,2, containing 0,15 M sodium chloride, 0,05 M phosphate buffer, and 0,09% sodium azide
  • Purification:
    Purified by multi-step chromatography

 

 

Isolated from bovine eye lens.