Mouse recombinant HSP60 (from E. coli)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Produced in E.coli.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Protein/peptide name:HSP60
- Endotoxin level:Low
- Purity:>90% (SDS-PAGE; Western blot)
- Formulation:Liquid. In 10 mm TRIS, pH 7,5, containing 150 mm sodium chloride, 1 mm EDTA, and 1 mm DTT
- Purification:Purified by multi-step chromatography
Specifications
Frequently Bought Together