Order Entry
Ireland
ContactUsLinkComponent
Calcineurin phosphatase assay kit
Calcineurin phosphatase assay kit
Catalog # ENZOBMLAK8040001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Calcineurin phosphatase assay kit
Catalog # ENZOBMLAK8040001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Calcineurin phosphatase assay kit
  • Seq:
    02
  • Storage conditions:
    –80 °C (long term)
  • Application:
    Colorimetric detection, HTS activity assay
  • Size:
    96 well

Specifications

About this item

This colorimetric kit is designed for inhibitor screening or kinetic assay of recombinant Calcineurin. Activity is determined using the RII phosphopeptide substrate, the most efficient and selective peptide known for calcineurin, and detection of free phosphate released is based on the classic malachite green assay.

  • Safe and non-radioactive assay
  • Convenient 1-step detection, no mixing needed
Delivery: Kit includes Recombinant calcineurin, RII phosphopeptide substrate, calmodulin, assay buffer, BIOMOL® GREEN detection reagent,
phosphate standard, microplate and detailed instructions.