Order Entry
Ireland
ContactUsLinkComponent
 
HSP70 ELISA Kit
Catalog # ENZOADIEKS700B
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
HSP70 ELISA Kit
Catalog # ENZOADIEKS700B
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Host:
  • Primary antibody reactivity:
  • Description:
    HSP70 ELISA Kit
  • Seq:
    02
  • Environmentally Preferable:
  • Sensitivity:
    200 pg/ml (range 780 - 50,000 pg/ml)

Specifications

About this item

Widely used ELISA HSP70 kit for chaperone and stress research.

For the quantitative determination of HSP70 in cell lysates and tissue of human, mouse, and rat origin. Prod. No. ADI-EKS-715 is recommended for the detection of Hsp70 in serum and plasma samples.

The HSP70 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4,5 hours. Absorbance is read at 450 nm. This kit is ideal for measuring intracellular HSP70 (HSP72). Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.

Ordering information: Contains Microtiter plate, 5X Extraction reagent, Standard, Sample diluent 2, Wash buffer concentrate, Antibody, Conjugate, TMB Substrate, Stop solution 2.