[pSer15]HSP27 (human), ELISA kit
Catalog # ENZOADI900170
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:1 KIT
- Conjugate ELISA:HRP
- Host:
- Primary antibody reactivity:Human
- Target protein:phospho-Ser15 HSP27
- Description:[pSer15]HSP27 (human), ELISA kit
- Seq:02
- Environmentally Preferable:
- Sample type:Cell lysates, plasma, and serum
- Detection method:Colorimetric
- Detection range:31,25 - 1000 pg/ml
- Sensitivity:10,15 pg/m
- Regulatory status:RUO
Specifications
About this item
Only commercially available ELISA kit for ultra-sensitive detection of human phospho-Ser15 HSP27 in cell lysates, plasma, and serum.
For the quantitative determination of human [pSer15]HSP27 in cell lysates, plasma, and serum.
The [pSer15]HSP27 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 27, Wash buffer concentrate, Standard, TMB Substrate, 5X Extraction reagent, Stop solution 2.