Order Entry
Ireland
ContactUsLinkComponent
IL-6 (mouse) ELISA kit
IL-6 (mouse) ELISA kit
Catalog # ENZOADI900045
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
IL-6 (mouse) ELISA kit
Catalog # ENZOADI900045
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Host:
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IL-6
  • Description:
    IL-6 (mouse) ELISA kit
  • Seq:
    02
  • Environmentally Preferable:
  • Sensitivity:
    1,01 pg/ml (range 7,81-1000 pg/ml)

Specifications

About this item

For the quantitative determination of mouse Il-6 in culture supernatants and serum. Cited sample type includes tissue.

The IL-6 (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours.

Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Wash buffer concentrate, Standard, TMB Substrate, Stop solution.