17β-Estradiol ELISA kit
Catalog # ENZOADI900008
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Competitive
- Host:
- Primary antibody reactivity:
- Target protein:17β-Estradiol
- Description:17β-Estradiol ELISA kit
- Seq:02
- Environmentally Preferable:
- Sensitivity:28,5 pg/ml (range 29.3 - 30000 pg/ml)
Specifications
About this item
For the quantitative determination of 17β-Estradiol in culture supernatants and saliva from any species. For serum and plasma use 89141-256.
The 17β-Estradiol EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405nm. This kit offers specific and save non-radioactive quantification of estradiol with ready-to-use liquid color-coded reagents to reduce error.
Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer 3, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-276 (5x96 wells)