Order Entry
Ireland
ContactUsLinkComponent
Anti-CRYAB Rabbit Polyclonal Antibody
Anti-CRYAB Rabbit Polyclonal Antibody
Supplier:  ENZO LIFE SCIENCES
New Product
Certificates
Anti-CRYAB Rabbit Polyclonal Antibody
Supplier:  ENZO LIFE SCIENCES
New Product

Specifications

  • Catalog No:
  • ENZOADISPA223F
  • ENZOADISPA223D
  • Antigen symbol:
  • CRYAB
  • CRYAB
  • Antigen name:
  • Crystallin Alpha B
  • Crystallin Alpha B
  • Antigen synonyms:
  • Heat-shock protein beta-5|HSPB5|CRYA2|Alpha Crystallin B chain|Alpha (B)-Crystallin|CryAB|Rosenthal fiber componet|CTPP2
  • Heat-shock protein beta-5|HSPB5|CRYA2|Alpha Crystallin B chain|Alpha (B)-Crystallin|CryAB|Rosenthal fiber componet|CTPP2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Form:
  • Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
  • Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
  • Purification:
  • Protein A affinity purified
  • Protein A affinity purified

Specifications

Frequently Bought Together