Order Entry
Ireland
ContactUsLinkComponent
 
Anti-FAS Mouse Monoclonal Antibody
  ENZOALX805038C100
Anti-FAS Mouse Monoclonal Antibody
  ENZOALX805038C100

 

  • Antibody type:
    Primary
  • Antigen name:
    Fatty Acid Synthase
  • Antigen symbol:
    FAS
  • Clonality:
    Monoclonal
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG3
  • Reactivity:
    Human
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 1% BSA. Contains no preservatives
  • Antigen synonyms:
    FASTM|APO-1|ALPS1A|TNFRSF6|CD95|APT1|FAS1
  • Concentration:
    1 mg/ml
  • Purification:
    ≥95% (SDS-PAGE)
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Host: Mouse, Isotype: IgG3

Recognizes human Fas.

Type: Primary
Antigen: FAS
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human