Order Entry
Ireland
ContactUsLinkComponent
Anti-GP9 Mouse Monoclonal Antibody [clone: Gi10]
Anti-GP9 Mouse Monoclonal Antibody [clone: Gi10]
Catalog # ENZOALX805023C100
Supplier:  ENZO LIFE SCIENCES
Anti-GP9 Mouse Monoclonal Antibody [clone: Gi10]
Catalog # ENZOALX805023C100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glycoprotein 9
  • Antigen symbol:
    GP9
  • Clonality:
    Monoclonal
  • Clone:
    Gi10
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Seq:
    02
  • Form:
    Liquid. 100 µg of purified antibody in 200 µl PBS containing 0,05% sodium azide
  • Antigen synonyms:
    glycoprotein 9|GP-IX|platelet glycoprotein IX|CD42a|Platelet GPIX|GPIX
  • Concentration:
    0,5 mg/ml
  • Purification:
    95% (SDS-PAGE)
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunoprecipitation

Recognizes the remnant moiety of the human CD42a/CD42b/CD42c/CD42d (GPIb/IX/V) complex.

Type: Primary
Antigen: CD42A
Clonality: Monoclonal
Clone: GI10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human