Anti-AKT Mouse Monoclonal Antibody
Catalog # ENZOALX804553C100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen symbol:AKT
- Clonality:Monoclonal
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Seq:02
- Form:Lyophilised from 1 ml of 2X PBS containing 0,09% sodium azide, PEG, and sucrose
- Purification:Thiophilic adsorption and size exclusion chromatography purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Host: Mouse, Isotype: IgG1
Recognizes a c-terminal epitope (aa 466-480) of phosphorylated and non-phosphorylated human, mouse, rat and dog Akt isoforms (see Technical Note below).
For ELISA, Flow Cytometry, Western Blot
Reconstitute with 1ml water (15 minutes at room temperature).
Ordering information: Positive control included.
Type: Primary
Antigen: AKT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human