Order Entry
Ireland
ContactUsLinkComponent
Anti-RANKL Mouse Monoclonal Antibody
Anti-RANKL Mouse Monoclonal Antibody
Catalog # ENZOALX804243C100
Supplier:  ENZO LIFE SCIENCES
Anti-RANKL Mouse Monoclonal Antibody
Catalog # ENZOALX804243C100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Receptor activator of nuclear factor kappa-B ligand
  • Antigen symbol:
    RANKL
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 0,05% BSA and 0,05% sodium azide.
  • Antigen synonyms:
    RANKLigand|Osteoprotegerin ligand|OPGL|sOdf|ODF|TRANCE|CD254|TNF-related activation-induced cytokine|RANKL|Receptor activator of nuclear factor kappa B ligand|hRANKL2|OPTB2
  • Concentration:
    0,5 mg/ml
  • Purification:
    Protein G-affinity purified
  • Pk:
    100 µG

Specifications

About this item

Host: Mouse, Isotype: IgG1 kappa

  • For Immunohistochemistry, Western Blot

Recognizes human and mouse RANKL. Detects a band of ~35-40kDa by Western blot.

Type: Primary
Antigen: RANKL
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human