Anti-ESR2 Rabbit Polyclonal Antibody
Catalog # ENZOALX210178C050
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:Estrogen Receptor 2
- Antigen symbol:ESR2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS containing 1 mg/ml BSA and 0,05% sodium azide.
- Antigen synonyms:ESR-BETA|ER-BETA|ESRB|Erb|NR3A2|ESTRB
- Concentration:1 mg/ml
- Purification:Epitope-affinity purified
- Pk:50 µG
Specifications
About this item
Host: Rabbit
- For Immunohistochemistry, Western Blot
Recognizes human and rat ERbeta. Detects a band of ~55kDa by Western blot. Does not cross-react with ERalpha.
Type: Primary
Antigen: ESR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human