Order Entry
Ireland
ContactUsLinkComponent
Anti-LEPR Rabbit Polyclonal Antibody
Anti-LEPR Rabbit Polyclonal Antibody
Catalog # ENZOALX210168R100
Supplier:  ENZO LIFE SCIENCES
Anti-LEPR Rabbit Polyclonal Antibody
Catalog # ENZOALX210168R100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Leptin Receptor
  • Antigen symbol:
    LEPR
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 0,05% sodium azide.
  • Antigen synonyms:
    Obr|Ob-R a/b/c/d/e|OB R gene related protein|Db|Leptin receptor.|Leptin receptor gene related protein|CD295|CD 295|OB-RGRP|HuB219|obl|LEPR|Fa|Leptin receptor precursor|OB receptor|LEPROT|LEPRD|OB-R|Leptin receptor fatty|LEPR_HUMAN|CD295 antigen|OB R|LEP R
  • Concentration:
    10 mg/ml
  • Purification:
    Purified IgG
  • Pk:
    100 µl

Specifications

About this item

Host: Rabbit

  • For Western Blot

Recognizes human, mouse and rat leptin receptor. Detects bands of ~100kDa and ~125kDa by Western blot, representing the short and long forms of leptin receptor. Also detects two unrelated bands of ~31kDa and ~35kDa.

Type: Primary
Antigen: LEPR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human