Specifications
- Antibody type:Primary
- Antigen symbol:DopamineR D1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS containing 50% glycerol and 0,01% sodium azide.
- Purification:Peptide affinity purified
- Pk:100 µG
Specifications
About this item
Dopamine is the predominant catecholamine neurotransmitter in the mammalian brain, where it regulates locomotor, emotional, memory, and dietary intake functions, as well as peripheral roles such as hormone secretion and renal function. Dopamine receptors (D1-D5) are members of the seven transmembrane domain containing G-protein coupled receptor (GPCR) family A, and are the target of several anti-psychosis and anti-Parkinsonian drugs. The D2, D3, and D4 dopamine receptors are encoded by homologous intron-containing genes, and signal via coupling of Gi/Go alpha G-protein subunits to inhibit adenylyl cyclase activity, as well as modulate cellular ion channel conductance of K+ and Ca2+. This is in contrast to D1 and D5 dopamine receptors, which signal via Gs alpha subunits to activate adenylyl cyclase activity. Coactivation of D1 and D2 receptors has been shown to activate a Gq mediated pathway, stimulating phospholipase C (PLC) and mobilizing intracellular calcium stores. D1 receptors were initially characterized in the striatum and retina and are expressed throughout the brain in the olfactory tubercle, islands of Calleja, entopeduncular nucleus, neocortex, hippocampus, and amygdala. Expression of the D2 receptor is found primarily in the brain (striatum, olfactory tubercle, and nucleus accumbens) and in the anterior and intermediate lobes of the pituitary.
Recognizes rat dopamine receptor D1. Predicted MW of ~49kDa. Higher MW species (~100kDa) and intermediates also observed by Western blot which reflect post-translational modification of the receptor. Predicted species reactivity based on epitope sequence homology: mouse, dog, bovine, wild boar.
For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Type: Primary
Antigen: DopamineR D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat