QUANTOFIX® Test Strips, Nickel
About this item
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. QUANTOFIX® test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
- Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips are for the rapid and reliable determination of Nickel in solutions. The easy dip-and-read-procedure provides a reliable result within 30 seconds.
2 Options Available Below
Product Details & Documents
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. QUANTOFIX® test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
- Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips are for the rapid and reliable determination of Nickel in solutions. The easy dip-and-read-procedure provides a reliable result within 30 seconds.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- MANA91327
- MANA91326
- Method:
- Carbonate hardness - Mixed indicator (bright green → blue);pH - mixed indicator (yellow → red);Total hardness - EDTA (green → red)
- Carbonate hardness - Mixed indicator (bright green → blue);pH - mixed indicator (yellow → red);Total hardness - EDTA (green → red)
- Parameters:
- All reagents included
- All reagents included
- Compliance:
- No hazardous material
- No hazardous material
- Contents:
- 25 test strips in a tube, instruction leaflet
- 100 test strips in a tube, instruction leaflet
- Instrument:
- No
- No
- Platform:
- Semi-quantitative test strips
- Semi-quantitative test strips
- Shelf life (months):
- 30 (from production)
- 30 (from production)
- Weight:
- 32,1 g
- 45,4 g
Specifications
Product Family Options
Product Information
- RangeChemical methodDimensionsTests per kitPkAvailabilityPrice
- QUANTOFIX® multistick for aquarium owners0 • 6,3 • 12,5 • 18,8 • 25,0 • 31,3 - °e; 0 • 4 • 8 • 13 • 19 • 25 - °e; 6,4 • 6,8 • 7,2 • 7,6 • 8,0 • 8,4Carbonate hardness: Reaction of an acid in the presence of a mix indicator; pH: pH indicator40×40×120 mm (packaging)25 Tests25Log in for availability1 * 25 items34,00 €Specifications:
More
Specifications
Cat. No.MANA91327MethodCarbonate hardness - Mixed indicator (bright green → blue);pH - mixed indicator (yellow → red);Total hardness - EDTA (green → red)ParametersAll reagents includedComplianceNo hazardous materialContents25 test strips in a tube, instruction leafletInstrumentNoPlatformSemi-quantitative test stripsShelf life (months)30 (from production)Weight32,1 g - QUANTOFIX® multistick for aquarium owners0 • 6,3 • 12,5 • 18,8 • 25,0 • 31,3 - °e; 0 • 4 • 8 • 13 • 19 • 25 - °e; 6,4 • 6,8 • 7,2 • 7,6 • 8,0 • 8,4Total hardness: color reaction with complex indicator40×40×120 mm (packaging)100 Tests100 TestsLog in for availability1 * 100 Tests67,80 €Specifications:
More
Specifications
Cat. No.MANA91326MethodCarbonate hardness - Mixed indicator (bright green → blue);pH - mixed indicator (yellow → red);Total hardness - EDTA (green → red)ParametersAll reagents includedComplianceNo hazardous materialContents100 test strips in a tube, instruction leafletInstrumentNoPlatformSemi-quantitative test stripsShelf life (months)30 (from production)Weight45,4 g