
Lanthanum(III) nitrate 0.1 M La(NO3)3 volumetric solution, for fluoride determination, Fluka™
Catalog # 35204-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.99 - 1.01
Specification Test Results
About this item
:
:324.92
:GHS05
Storage Temperature:Ambient
CAS Number:10099-59-9
Formula:LaN₃O₉
MDL Number:MFCD00011067