Order Entry
Ireland
ContactUsLinkComponent
Combined fridge freezers, BioFresh, CBNesf series
Combined fridge freezers, BioFresh, CBNesf series
  LIEBCBNESF5113
 :  Liebherr
Combined fridge freezers, BioFresh, CBNesf series
  LIEBCBNESF5113
 :  Liebherr
  • W×D×H ext.:
    750×630×2020 mm
  • Capacity:
    300 + 115 l
  • Description:
    Combined refrigerator/freezer, silver colour coated steel, CBNesf 5113, with fan in refrigerator, right interchangeable door hinges
  • Seq:
    02
  • Model:
    CBNesf 5113 Comfort Biofresh NoFrost
  • Cat. No.:
    LIEBCBNESF5113
  • Alarm:
    audible, optical and acoustical
  • Defrost:
    automatic
  • Display:
    LED temperature display, internal analog
  • Fans:
    Fan in refrigerator
  • Material:
    Steel
  • Shelves/drawers:
    7 glass with plastic frame shelves (refrigerator), 3 glass drawers (freezer)
  • Specifications:
    Stainless steel slimline door handle with integrated opening mechanism
  • Energy class:
    A+
  • Pk:
    1
These combined fridge freezers with biofresh and NoFrost offer more interior capacity for food storage, economy and energy efficiency.
  • Quiet operation, ease-of-use and exceptional design
  • Sturdy, secure handles have an elegant, premium quality appearance
  • LED ceiling light
  • High-performance power cooling system
These freezers feature stainless steel door with SmartSteel and aluminium door handle with integrated opening mechanism. The NoFrost system provides rapid, fan-assisted, frost-free freezing for the safe, longterm storage of food. The NoFrost technology creates a much larger storage capacity and keeps the freezer constantly frost-free. The BioFresh safes pull out on smooth-running telescopic rails and can easily be removed once the door is open to an angle of 90°
Accurate and independent temperature control of the fridge and freezer compartments is facilitated by DuoCooling: two separately adjustable cooling circuits. There is no air exchange between the fridge and freezer compartment, so that odours cannot be transferred and the stored food is prevented from drying out.